Assay Details
| Gene Symbol: TP53INP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634952.3 | XP_005635009.1 | 2 - 3 | 1184 | 115 | |
| XM_005634953.3 | XP_005635010.1 | 2 - 3 | 342 | 115 | ||
| XM_005634955.3 | XP_005635012.1 | 2 - 3 | 582 | 115 | ||
| XM_022409384.1 | XP_022265092.1 | 3 - 4 | 1294 | 115 | ||
| XM_022409385.1 | XP_022265093.1 | 2 - 3 | 590 | 115 | ||
| XM_022409386.1 | XP_022265094.1 | 2 - 3 | 520 | 115 | ||
More Information
Set Membership: |
|

TP53INP2
Genomic Map