Assay Details
| Gene Symbol: RAB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003153.1 | NP_001003153.1 | 4 - 5 | 436 | 74 | |
| GenBank mRNA | X56384.1 | - | 436 | 74 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAB1A
Genomic Map