Assay Details
| Gene Symbol: TRAPPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628951.3 | XP_005629008.1 | 4 - 5 | 640 | 147 | |
| XM_539594.6 | XP_539594.2 | 4 - 5 | 545 | 147 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TRAPPC3
Genomic Map