Assay Details
| Gene Symbol: NSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629854.2 | XP_005629911.1 | 3276 | 66 | ||
| XM_005629856.2 | XP_005629913.1 | 3276 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629854.2 | XP_005629911.1 | 3276 | 66 | ||
| XM_005629856.2 | XP_005629913.1 | 3276 | 66 | |||
Set Membership: |
|