Assay Details
| Gene Symbol: ANGPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005754.2 | NP_001005754.1 | 1052 | 119 | ||
| XM_005627803.3 | XP_005627860.1 | 1524 | 119 | |||
| XM_022426493.1 | XP_022282201.1 | 1527 | 119 | |||
| GenBank mRNA | AB192412.2 | - | 1052 | 119 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ANGPT1
Genomic Map