Assay Details
| Gene Symbol: LOC611401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635169.3 | XP_005635226.1 | 5 - 6 | 691 | 71 | |
| XM_849066.6 | XP_854159.3 | 5 - 6 | 779 | 71 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC611401
Genomic Map