Assay Details
| Gene Symbol: KYAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621957.1 | XP_005622014.1 | 9 - 10 | 986 | 136 | |
| XM_005621958.3 | XP_005622015.1 | 9 - 10 | 1137 | 136 | ||
| XM_537084.6 | XP_537084.1 | 9 - 10 | 908 | 136 | ||
More Information
Set Membership: |
|

KYAT3
Genomic Map