Assay Details
| Gene Symbol: RGS7BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617437.3 | XP_005617494.2 | 4 - 5 | 532 | 86 | |
| XM_846835.5 | XP_851928.1 | 4 - 5 | 641 | 86 | ||
More Information
Set Membership: |
|

RGS7BP
Genomic Map