Assay Details
| Gene Symbol: FYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627719.2 | XP_005627776.1 | 13 - 14 | 1841 | 91 | |
| XM_849374.4 | XP_854467.1 | 13 - 14 | 1850 | 91 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FYN
Genomic Map