Assay Details
| Gene Symbol: SRSF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623582.3 | XP_005623639.1 | 5 - 6 | 559 | 102 | |
| XM_014116009.2 | XP_013971484.1 | 5 - 6 | 522 | 102 | ||
| XM_014116010.2 | XP_013971485.1 | 6 - 7 | 748 | 102 | ||
| XM_014116011.2 | XP_013971486.1 | 5 - 6 | 556 | 102 | ||
| XM_022422168.1 | XP_022277876.1 | 5 - 6 | 562 | 102 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SRSF5
Genomic Map