Assay Details
| Gene Symbol: PKNOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014113315.2 | XP_013968790.1 | 2067 | 57 | ||
| XM_014113316.2 | XP_013968791.1 | 2057 | 57 | |||
| XM_014113318.2 | XP_013968793.1 | 1058 | 57 | |||
| XM_022418131.1 | XP_022273839.1 | 1951 | 57 | |||
| XM_022418132.1 | XP_022273840.1 | 2024 | 57 | |||
| XM_022418133.1 | XP_022273841.1 | 2140 | 57 | |||
| XM_022418134.1 | XP_022273842.1 | 1844 | 57 | |||
| XM_022418135.1 | XP_022273843.1 | 1842 | 57 | |||
| XM_536535.6 | XP_536535.4 | 1319 | 57 | |||
More Information
Set Membership: |
|

PKNOX2
Genomic Map