Assay Details
| Gene Symbol: DCAF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022425623.1 | XP_022281331.1 | 2 - 3 | 698 | 76 | |
| XM_538739.5 | XP_538739.2 | 2 - 3 | 698 | 76 | ||
More Information
Set Membership: |
|

DCAF10
Genomic Map