Assay Details
Gene Symbol: KEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005632897.3 | XP_005632954.1 | 3 - 4 | 1502 | 62 | |
XM_533917.6 | XP_533917.2 | 3 - 4 | 1526 | 62 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
