Assay Details
| Gene Symbol: CRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435202.4 | XP_003435250.1 | 1 - 2 | 393 | 131 | |
| XM_003435203.4 | XP_003435251.1 | 1 - 2 | 393 | 131 | ||
| XM_022423445.1 | XP_022279153.1 | 1 - 2 | 396 | 131 | ||
More Information
Set Membership: |
|

CRK
Genomic Map