Assay Details
| Gene Symbol: MATR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205258.1 | NP_001192187.1 | 14 - 15 | 2716 | 127 | |
| XM_005626403.3 | XP_005626460.1 | 14 - 15 | 2950 | 127 | ||
| XM_005626404.3 | XP_005626461.1 | 15 - 16 | 3031 | 127 | ||
| XM_005626405.3 | XP_005626462.1 | 15 - 16 | 3022 | 127 | ||
| XM_005626406.3 | XP_005626463.1 | 14 - 15 | 2914 | 127 | ||
| XM_014117660.2 | XP_013973135.1 | 17 - 18 | 3100 | 127 | ||
| GenBank mRNA | DN368188.1 | - | 2 - 3 | 234 | 127 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MATR3
Genomic Map