Assay Details
| Gene Symbol: GSK3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639536.3 | XP_005639593.1 | 4 - 5 | 685 | 81 | |
| XM_535751.6 | XP_535751.2 | 4 - 5 | 685 | 81 | ||
| XM_851518.5 | XP_856611.1 | 4 - 5 | 685 | 81 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GSK3B
Genomic Map