Assay Details
| Gene Symbol: CNPPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640194.4 | XP_003640242.1 | 5 - 6 | 686 | 72 | |
| XM_005640678.3 | XP_005640735.1 | 5 - 6 | 868 | 72 | ||
| XM_014111054.2 | XP_013966529.1 | 6 - 7 | 748 | 72 | ||
| XM_022415261.1 | XP_022270969.1 | 6 - 7 | 923 | 72 | ||
| XM_022415262.1 | XP_022270970.1 | 7 - 8 | 780 | 72 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CNPPD1
Genomic Map