Assay Details
| Gene Symbol: HNRNPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109983.2 | XP_013965458.1 | 3 - 4 | 877 | 105 | |
| XM_535627.6 | XP_535627.2 | 3 - 4 | 878 | 105 | ||
| XM_843857.5 | XP_848950.1 | 4 - 5 | 934 | 105 | ||
| XM_852864.5 | XP_857957.1 | 4 - 5 | 935 | 105 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HNRNPD
Genomic Map