Assay Details
| Gene Symbol: SMNDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637715.3 | XP_005637772.1 | 4 - 5 | 697 | 64 | |
| XM_005637716.2 | XP_005637773.1 | 4 - 5 | 612 | 64 | ||
| XM_535011.5 | XP_535011.2 | 4 - 5 | 549 | 64 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SMNDC1
Genomic Map