Assay Details
| Gene Symbol: ENO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619919.3 | XP_005619976.1 | 7 - 8 | 827 | 58 | |
| XM_022418447.1 | XP_022274155.1 | 8 - 9 | 913 | 58 | ||
| XM_536606.6 | XP_536606.4 | 7 - 8 | 809 | 58 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ENO3
Genomic Map