Assay Details
| Gene Symbol: PIGH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623556.3 | XP_005623613.1 | 1 - 2 | 278 | 56 | |
| XM_005623557.3 | XP_005623614.1 | 1 - 2 | 279 | 56 | ||
| XM_849246.5 | XP_854339.2 | 1 - 2 | 274 | 56 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PIGH
Genomic Map