Assay Details
| Gene Symbol: PRKCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022422096.1 | XP_022277804.1 | 8 - 9 | 855 | 116 | |
| XM_022422097.1 | XP_022277805.1 | 8 - 9 | 802 | 116 | ||
| XM_547844.6 | XP_547844.2 | 8 - 9 | 1483 | 116 | ||
More Information
Set Membership: |
|

PRKCH
Genomic Map