Assay Details
Gene Symbol: DDX39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014377.1 | NP_001014399.1 | 7 - 8 | 979 | 93 | |
XM_022425718.1 | XP_022281426.1 | 8 - 9 | 1133 | 93 | ||
XM_022425719.1 | XP_022281427.1 | 8 - 9 | 1251 | 93 | ||
XM_022425720.1 | XP_022281428.1 | 8 - 9 | 1133 | 93 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
