Assay Details
| Gene Symbol: CHIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628193.3 | XP_005628250.1 | 3 - 4 | 2455 | 107 | |
| XM_022426995.1 | XP_022282703.1 | 3 - 4 | 318 | 107 | ||
| XM_022426996.1 | XP_022282704.1 | 4 - 5 | 2649 | 107 | ||
More Information
Set Membership: |
|

CHIC2
Genomic Map