Assay Details
| Gene Symbol: VPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639850.4 | XP_003639898.1 | 7 - 8 | 846 | 79 | |
| XM_014107172.2 | XP_013962647.1 | 8 - 9 | 870 | 79 | ||
| XM_022408918.1 | XP_022264626.1 | 5 - 6 | 555 | 79 | ||
More Information
Set Membership: |
|

VPS16
Genomic Map