Assay Details
| Gene Symbol: SZRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617952.2 | XP_005618009.1 | 1 - 2 | 132 | 121 | |
| XM_846908.4 | XP_852001.2 | 1 - 2 | 132 | 124 | ||
More Information
Set Membership: |
|

SZRD1
Genomic Map