Assay Details
| Gene Symbol: NRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434488.4 | XP_003434536.1 | 5 - 6 | 1210 | 69 | |
| XM_005619098.3 | XP_005619155.1 | 5 - 6 | 1207 | 69 | ||
| XM_014112927.2 | XP_013968402.1 | 5 - 6 | 1215 | 69 | ||
| XM_014112928.2 | XP_013968403.1 | 5 - 6 | 1214 | 69 | ||
| XM_014112929.2 | XP_013968404.1 | 5 - 6 | 1212 | 69 | ||
| XM_014112930.2 | XP_013968405.1 | 4 - 5 | 1188 | 69 | ||
| XM_014112931.2 | XP_013968406.1 | 5 - 6 | 1212 | 69 | ||
More Information
Set Membership: |
|

NRG3
Genomic Map