Assay Details
| Gene Symbol: AQP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003130.1 | NP_001003130.1 | 1 - 2 | 421 | 65 | |
| GenBank mRNA | AB011373.1 | - | 1 - 2 | 421 | 65 | |
| AF180528.1 | - | 1 - 2 | 69 | 65 | ||
More Information
Set Membership: |
|

AQP1
Genomic Map