Assay Details
| Gene Symbol: CES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003085.1 | NP_001003085.1 | 8 - 9 | 946 | 88 | |
| GenBank mRNA | AB023629.1 | - | 8 - 9 | 946 | 88 | |
More Information
Set Membership: |
|

CES1
Genomic Map