Assay Details
| Gene Symbol: MT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003150.1 | NP_001003150.1 | 2 - 3 | 138 | 88 | |
| GenBank mRNA | AB028041.1 | - | 2 - 3 | 138 | 88 | |
More Information
Set Membership: |
|

MT4
Genomic Map