Assay Details
| Gene Symbol: CA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002999.1 | NP_001002999.1 | 743 | 89 | ||
| GenBank mRNA | AB080972.1 | - | 743 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002999.1 | NP_001002999.1 | 743 | 89 | ||
| GenBank mRNA | AB080972.1 | - | 743 | 89 | ||
Set Membership: |
|