Assay Details
| Gene Symbol: REG3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002945.3 | NP_001002945.2 | 3 - 4 | 257 | 81 | |
| XM_005630062.3 | XP_005630119.1 | 3 - 4 | 233 | 81 | ||
| XM_022404313.1 | XP_022260021.1 | 3 - 4 | 233 | 81 | ||
| GenBank mRNA | GR887671.1 | - | 3 - 4 | 257 | 81 | |
| Gene Symbol: LOC100687463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256535.2 | NP_001243464.1 | 2 - 3 | 214 | 81 | |
| XM_014120197.2 | XP_013975672.1 | 3 - 4 | 248 | 81 | ||
| XM_022404312.1 | XP_022260020.1 | 3 - 4 | 233 | 81 | ||
More Information
Set Membership: |
|

REG3A
Genomic Map