Assay Details
| Gene Symbol: CELA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003007.2 | NP_001003007.2 | 4 - 5 | 373 | 57 | |
| GenBank mRNA | AB100595.1 | - | 3 - 4 | 327 | 57 | |
| DN357055.1 | - | 4 - 5 | 394 | 57 | ||
| DN357380.1 | - | 4 - 5 | 372 | 57 | ||
| DN359987.1 | - | 4 - 5 | 343 | 57 | ||
More Information
Set Membership: |
|

CELA1
Genomic Map