Assay Details
| Gene Symbol: TLR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002998.1 | NP_001002998.1 | 1 - 2 | 64 | 87 | |
| GenBank mRNA | AB104899.3 | - | 1 - 2 | 64 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001002998.1 transcripts.
Set Membership: |
|

TLR9
Genomic Map