Assay Details
| Gene Symbol: CCL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003968.1 | NP_001003968.1 | 1 - 2 | 68 | 87 | |
| XM_022424909.1 | XP_022280617.1 | 2 - 3 | 321 | 87 | ||
| GenBank mRNA | AB162850.1 | - | 1 - 2 | 68 | 87 | |
More Information
Set Membership: |
|

CCL27
Genomic Map