Assay Details
| Gene Symbol: CCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005255.1 | NP_001005255.1 | 1 - 2 | 75 | 61 | |
| GenBank mRNA | AB163436.1 | - | 1 - 2 | 75 | 61 | |
| AY675234.1 | - | 1 - 2 | 75 | 61 | ||
More Information
Set Membership: |
|

CCL8
Genomic Map