Assay Details
| Gene Symbol: HTR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005869.1 | NP_001005869.1 | 608 | 74 | ||
| GenBank mRNA | AB193092.1 | - | 608 | 74 | ||
| AY832858.1 | - | 608 | 74 | |||
| Y16134.1 | - | 252 | 74 | |||
More Information
Set Membership: |
|

HTR2A
Genomic Map