Assay Details
| Gene Symbol: CCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038606.2 | NP_001033695.2 | 2 | 1318 | 84 | |
| XM_005632330.3 | XP_005632387.1 | 2 | 1377 | 84 | ||
| GenBank mRNA | AB219241.1 | - | 1 | 1009 | 84 | |
More Information
Set Membership: |
|

CCR1
Genomic Map