Assay Details
| Gene Symbol: OBP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003190.1 | NP_001003190.1 | 3 - 4 | 296 | 63 | |
| GenBank mRNA | AF027177.1 | - | 3 - 4 | 296 | 63 | |
More Information
Set Membership: |
|

OBP2B
Genomic Map