Assay Details
| Gene Symbol: OPN1LW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197072.1 | NP_001184001.1 | 2 - 3 | 411 | 77 | |
| GenBank mRNA | AF031533.1 | - | 1 - 2 | 270 | 77 | |
More Information
Set Membership: |
|

OPN1LW
Genomic Map