Assay Details
| Gene Symbol: KCND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630867.3 | XP_005630924.1 | 7 | 1884 | 65 | |
| XM_005630868.3 | XP_005630925.1 | 6 | 1829 | 65 | ||
| XM_540343.6 | XP_540343.2 | 6 | 1825 | 65 | ||
| XM_845974.5 | XP_851067.1 | 7 | 1880 | 65 | ||
| GenBank mRNA | AF049887.1 | - | 1 | 44 | 65 | |
More Information
Set Membership: |
|

KCND3
Genomic Map