Assay Details
| Gene Symbol: TP53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003210.1 | NP_001003210.1 | 1 - 2 | 95 | 66 | |
| GenBank mRNA | AB537893.1 | - | 2 - 3 | 235 | 66 | |
| AF060514.1 | - | 1 - 2 | 95 | 66 | ||
| KP279761.1 | - | 1 - 2 | 79 | 66 | ||
| KP279762.1 | - | 1 - 2 | 79 | 66 | ||
| KP279763.1 | - | 1 - 2 | 79 | 66 | ||
More Information
Set Membership: |
|

TP53
Genomic Map