Assay Details
| Gene Symbol: ADRA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195843.1 | NP_001182772.1 | 1 | 831 | 56 | |
| XM_005635607.3 | XP_005635664.1 | 3 | 1888 | 56 | ||
| XM_022409542.1 | XP_022265250.1 | 2 | 2138 | 56 | ||
| XM_022409544.1 | XP_022265252.1 | 2 | 2142 | 56 | ||
| XM_022409545.1 | XP_022265253.1 | 2 | 2140 | 56 | ||
| XM_022409546.1 | XP_022265254.1 | 2 | 2149 | 56 | ||
| GenBank mRNA | AF068283.1 | - | 1 | 832 | 56 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ADRA1A
Genomic Map