Assay Details
| Gene Symbol: GH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003168.1 | NP_001003168.1 | 225 | 71 | ||
| GenBank mRNA | AF069071.1 | - | 225 | 71 | ||
| S78184.1 | - | 61 | 71 | |||
| Z23067.1 | - | 171 | 71 | |||
More Information
Set Membership: |
|

GH1
Genomic Map