Assay Details
| Gene Symbol: CD86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003146.2 | NP_001003146.1 | 6 - 7 | 918 | 68 | |
| XM_014110137.2 | XP_013965612.1 | 6 - 7 | 1070 | 68 | ||
| XM_014110138.2 | XP_013965613.1 | 5 - 6 | 925 | 68 | ||
| GenBank mRNA | AF106826.1 | - | 6 - 7 | 906 | 68 | |
| AF106827.1 | - | 5 - 6 | 760 | 68 | ||
More Information
Set Membership: |
|

CD86
Genomic Map