Assay Details
| Gene Symbol: GHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003123.1 | NP_001003123.1 | 3 - 4 | 284 | 68 | |
| XM_005618865.3 | XP_005618922.1 | 4 - 5 | 303 | 68 | ||
| XM_014112763.1 | XP_013968238.1 | 4 - 5 | 303 | 68 | ||
| XM_014112764.1 | XP_013968239.1 | 4 - 5 | 301 | 68 | ||
| XM_022417326.1 | XP_022273034.1 | 3 - 4 | 237 | 68 | ||
| GenBank mRNA | AF133835.1 | - | 3 - 4 | 284 | 68 | |
More Information
Set Membership: |
|

GHR
Genomic Map