Assay Details
| Gene Symbol: SRD5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287064.1 | NP_001273993.1 | 2 - 3 | 446 | 99 | |
| GenBank mRNA | AF136721.1 | - | 1 - 2 | 162 | 99 | |
More Information
Set Membership: |
|

SRD5A2
Genomic Map