Assay Details
Gene Symbol: KCNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003113.1 | NP_001003113.1 | 1 - 2 | 575 | 81 | |
XM_022407415.1 | XP_022263123.1 | 1 - 2 | 1035 | 81 | ||
XM_022407416.1 | XP_022263124.1 | 1 - 2 | 1036 | 81 | ||
XM_022407417.1 | XP_022263125.1 | 1 - 2 | 1036 | 81 | ||
GenBank mRNA | AF153198.1 | - | 1 - 2 | 575 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
