Assay Details
| Gene Symbol: BGLAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115322.2 | XP_013970797.1 | 4 | 307 | 54 | |
| GenBank mRNA | AF205942.1 | - | 1 | 52 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: BGLAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115322.2 | XP_013970797.1 | 4 | 307 | 54 | |
| GenBank mRNA | AF205942.1 | - | 1 | 52 | 54 | |
Set Membership: |
|