Assay Details
| Gene Symbol: ATP6V0E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003128.1 | NP_001003128.1 | 2 - 3 | 191 | 66 | |
| GenBank mRNA | AF258614.1 | - | 2 - 3 | 191 | 66 | |
| AF343440.1 | - | 2 - 3 | 181 | 66 | ||
More Information
Set Membership: |
|

ATP6V0E1
Genomic Map